Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Endothelin 1, Human and Porcine, Polypeptide, Calbiochem

Supplier:  MilliporeSigma™ 052338000.5MG

 View more versions of this product

Catalog No. 05-233-80005M


Add to Cart

Description

Description

A 21-amino acid polypeptide with potent vasoconstrictive action. Produces convulsions, arterial hypertension, and metabolic activation of periventricular white and gray matter. Its effects are mediated via L-type Ca2+ channels. Reported to activate phospholipase C in fibroblasts expressing ETA receptors.
Specifications

Specifications

Human, Porcine
≥96%
White to Off-white
Solid
Endothelin 1
RUO - research use only
HPLC
117399-94-7
Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 2 weeks at −20°C.
Supplied as an acetate salt.
0.5 mg
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit