Learn More
MilliporeSigma™ Endothelin 3, Human and Rat, peptide, Calbiochem
Supplier:  MilliporeSigma™ 052338010.5MG
Catalog No. 05-233-80105M
Add to Cart
Description
Description
Attenuates thrombin-evoked aggregation of human platelets. A potent stimulator of interleukin-6 production by endothelial cell lines. Reported to activate phospholipase C in fibroblasts expressing ETA receptors.
Specifications
Specifications
Human, Rat
≥97%
Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 1 month at −20°C.
Supplied as a trifluoroacetate salt. Sold on the basis of peptide content.
0.5 mg
HPLC
117399-93-6
Solid
Endothelin 3
RUO - research use only
Product Suggestions
Product Suggestions