Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Enzyme Free Cell Dissociation Solution PBS Based (1x)
Click to view available options
Quantity:
100 mL
500 mL
Description
- Works significantly faster than 0.5mM EDTA
- Preserves structural and functional integrity of cell surface proteins
- Does not have cytotoxic effects sometimes associated with 0.5 mM EDTA
- Used with cells that are sub-confluent (60-80%)

Specifications
Specifications
Content And Storage | Store at room temperature or at 4°C. |
Concentration | 1 X |
Form | Liquid |
Product Type | Cell Culture Dissociation Reagent |
Sterility | Sterile |
Formulation | Chelating and stabilizing agents |
For Use With (Application) | Cell Dissociation |
Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction