Learn More
MilliporeSigma™ Epidermal Growth Factor (EGF), Culture Grade
This Epidermal growth factor (EGF), culture grade product is available in a 100μg format.
Manufacturer:  MilliporeSigma™ 01101
Catalog No. 01-101MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide.
Specifications
Specifications
Lyophilized
Cell Culture
100μg