Learn More
MilliporeSigma™ Epidermal Growth Factor (EGF), Receptor Grade
This Epidermal growth factor (EGF), receptor grade product is available in a 100μg format.
Manufacturer:  MilliporeSigma™ 01102
Catalog No. 01-102MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide.
Specifications
Specifications
Greater than or equal to 99% by SDS-PAGE/Coomassie blue staining
Lyophilized
100μg
Cell Culture