Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ESGRO Complete™ Gelatin Solution

For stem cell culture

$42.25

Specifications

For Use With (Application) Stem Cell Culture
Format 0.1% gelatin solution sourced from porcine
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
SF008
View Documents
MilliporeSigma™
SF008
500mL
Each for $42.25
Add to cart
 
Description

Description

Specifications

Specifications

Stem Cell Culture
0.1% gelatin solution sourced from porcine
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit