Learn More
Formaldehyde Concentrate, MilliporeSigma™
Catalog Number MR0466901
Click to view available options
Quantity:
Each
Prepares buffered neutral formalin, 10%
Contain buffer concentrate (x10) and formaldehyde solution 37% W/V
  • Packaged in Poly bottle
Quantity Each