Learn More
MilliporeSigma™ EXtrelut NT Prepacked Column: Filters
Spare filters for EXtrelut™ NT prepacked glass columns
Manufacturer:  MilliporeSigma™ 1.14237.0001
 View more versions of this product
Catalog No. M1142370001
Add to cart
Description
Description
Extrelut™ NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut™ NT1 can take up a maximum of one ml, Extrelut™ NT3 threemL and Extrelut™ NT20 up to twentymL of sample.
Specifications
Specifications
EXtrelut NT3 Prepacked Glass Columns
100 Units
Replacement Filter