Learn More
MilliporeSigma EXtrelut NT Prepacked Column: Filters
Spare filters for EXtrelut NT prepacked glass columns
$119.00 - $198.60
Catalog Number | Mfr. No. | For Use With | Packaging Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With | Packaging Quantity | Price | Quantity & Availability | ||||
M1142360001
|
MilliporeSigma
1.14236.0001 |
EXtrelut NT1 Prepacked Glass Columns | 100 Units |
Each for $168.30
|
|
M1142370001
|
MilliporeSigma
1.14237.0001 |
EXtrelut NT3 Prepacked Glass Columns | 100 Units |
Each for $119.00
|
|
M1145670001
|
MilliporeSigma
1.14567.0001 |
EXtrelut NT 20 HDPE Prepacked Columns | 50 Units |
Each for $198.60
|
|
Description
Extrelut NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut NT1 can take up a maximum of one ml, Extrelut NT3 threemL and Extrelut NT20 up to twentymL of sample.Specifications
Replacement Filter |