Learn More
MilliporeSigma™ Fast-Trap™ Adenovirus and Lentivirus Purification and Concentration Kits
Quick, easy membrane-based method for laboratory-scale virus purification
Manufacturer:  MilliporeSigma™ FTLV00003
 View more versions of this product
Catalog No. FTLV00003
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
These kits permit quick purification of infectious agents which should be handled under BSL2 safety precautions. Wear hand, eye, face, and body personal protective equipment (PPE) when using this kit. All virus purification steps (except water bath incubation and centrifugation) should be conducted in a biosafety cabinet.
Includes: Three Fast-Trap virus filter units, three Steriflip-HV filter units, 20mL 10X binding buffer, 100mL equilibration buffer, 200mL wash buffer, 21mL elution buffer, three Amicon™ Ultra-4 filter units, test tube stand
Description
Description
Fast-Trap Purification
  • Saves time—protocol can be completed in less than two hours
  • High yield—recover up to 70% of virus particles
  • Dependable—simple protocol ensures quality results
  • Easy—Steriflip™ filters eliminate messy purification steps and expensive equipment
  • Safe—use a closed vacuum device to eliminate potential spills
Specifications
Specifications
2 x 108 virus particles
Adenovirus purification and concentration
<2 hr.
3.0
Fast trap adenovirus purification and concentration kit
Three fast trap virus filter units, three steriflip HV filter units, 20mL 10X Binding Buffer, 100mL Equilibration Buffer, 200mL Wash Buffer, 21mL Elution Buffer, three Amicon ultra 4 filter units, test tube stand
Room Temperature
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For laboratory use only; not for diagnostic or in vivo use.