EMD Millipore™ Fibrinogen, Human Plasma, Protein Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: EMD Millipore 3415761GM

 View more versions of this product

Catalog No. 34-157-61GM

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Preparation Method Sterile dH2O
    Purification Method Homogeneous by SDS-PAGE
    Quantity 1g
    Recombinant No
    Immunogen Fibrinogen
    Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Do not store solutions on ice; precipitation will occur. Stock solutions are stable for up to 6 months at −70°C. Avoid freeze-thaw cycles.
    Regulatory Status RUO
    Format Lyophilized
    Formulation Lyophilized from 20mM sodium citrate-HCl, pH 7.4.
    Host Species Human plasma

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit