Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Fibroblast Growth Factor-Basic Human Recombinant Protein Available on GSA/VA Contract for Federal Government customers only.

$322.00 - $1641.00

Specifications

Cross Reactivity Human
Purity Greater than 95% by SDS-PAGE
Recombinant Yes
Name Fibroblast Growth Factor-basic (FGF-b)
For Use With (Application) in vitro applications
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
GF003AFMG
View Documents
MilliporeSigma™
GF003AFMG
1mg
Each for $1,641.00
Add to cart
 
GF003 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
GF003
50μg
Each for $322.00
Add to cart
 
Description

Description

Specifications

Specifications

Human
Yes
in vitro applications
Greater than 95% by SDS-PAGE
Fibroblast Growth Factor-basic (FGF-b)
Expressed in E. coli
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit