Learn More
MilliporeSigma&trade FlowCellect™ Oxidative Stress Characterization Kit Available on GSA/VA Contract for Federal Government customers only.
Oxidative stress has been found to play a key role in a number of pathological disorders.
Manufacturer:  MilliporeSigma&trade FCCH025111
 View more versions of this product
Catalog No. FCCH025111
Add to cart
Description
Description
Oxidative stress has been found to play a key role in a number of pathological disorders. To aid researchers studying this phenomenon Millipore has developed the FlowCellect™ Oxidative Stress Characterization Kit. This kit contains the chemical and immunological reagents necessary to detect carbonyl groups introduced onto proteins by reactive oxygen species (ROS). The test method involves derivatization of protein carbonyl groups with 2,4-dinitrophenylhydrazine (DNPH). This chemical reaction results in proteins being covalently coupled to DNP at their carbonyl sites. The DNP-derivatized proteins are then incubated with a FITC conjugated monoclonal antibody that specifically binds to the DNP moiety. Subsequent detection with a bench-top Guava EasyCyte™ Plus flow cytometer allows quantification of the extent of oxidative stress within cells. Although the assay and all of the kit components were optimized using a Guava instrument, any flow cytometer can be utilized provided it contains a blue laser (488 nm) light source.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Flow Cytometry
FlowCellect oxidative stress characterization kit
Upon arrival the FITC conjugated anti-DNP antibody should be transferred to a -20°C freezer. The remaining kit components should be stored at 2-8°C
User manual, anti-DNP -FITC, derivatization control solution, 5X assay buffer, DNPH solution, 10X wash buffer
25 tests