EMD Millipore™ Gonadotropin, Pregnant Mare Serum, Protein Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: EMD Millipore 3672225000IU

 View more versions of this product

Catalog No. 36-722-25000I

  Check Availability
Add to Cart

    Specifications

    Preparation Method H2O
    Quantity 5000IU
    Recombinant No
    Immunogen Gonadotropin
    Storage Requirements Following reconstitution store in the refrigerator (4°C). Stock solutions are stable for up to 1 week at 4°C.
    Regulatory Status RUO
    Format White solid
    Host Species Pregnant Mare Serum

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit