EMD Millipore HARLECO™ Fuchsin Carbol Solution Available on GSA/VA Contract for Federal Government customers only.

For acid-fast staining of micro-organisms

Manufacturer: EMD Millipore 6051X75

 View more versions of this product

Catalog No. 23-749981

  Check Availability
Add to Cart

    Specifications

    Stain Type Carbolfuchsin
    Type Ziehl Neelsen Formula
    Quantity 1L
    Format Bottle
    For Use With (Application) Acid fast staining of microorganisms

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit