EMD Millipore Harleco™ Fucillo Wright Giemsa Solution Available on GSA/VA Contract for Federal Government customers only.

For use with the MIDAS™ III Automated Stainer

Manufacturer: EMD Millipore 6457195

 View more versions of this product

Catalog No. 23-750001

  Check Availability
Add to Cart

    Specifications

    Quantity 500mL
    Boiling Point 64.5°C
    Format Liquid
    Type Fucillo Wright Giemsa Solution

    • Polychromatic stain for peripheral blood and bone marrow smears
    • Used for red blood cell morphology and white blood cell differentiation

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit