EMD Millipore HARLECO™ Gram Stain Reagents Available on GSA/VA Contract for Federal Government customers only.

Complete set contains all reagents needed for Gram staining, or choose reagents individually

Manufacturer: EMD Millipore 65092B95

 View more versions of this product

Catalog No. 23-750026

  Check Availability
Add to Cart

    Specifications

    Stain Type Safranin
    Type Safranin Stain Reagent
    Quantity 4 x 500mL
    Format Bottle
    For Use With (Application) Stains gram-negative bacteria red

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit