Learn More
EMD Millipore Harleco™ Histology/Papanicolaou Stains: Aqueous Eosin Y Solution Available on GSA/VA Contract for Federal Government customers only.
Harleco Histology/Papanicolaou Stains
Manufacturer:  EMD Millipore 59275
Catalog No. 23-749978
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Aqueous Eosin Y Solution
1000mL
Cytoplasmic counterstain for Hemotoxylin
Accessories