EMD Millipore Harleco™ Histology/Papanicolaou Stains: EA-36 Papanicolaou Staining Solution Available on GSA/VA Contract for Federal Government customers only.
Harleco™ Histology/Papanicolaou Stains
Manufacturer: EMD Millipore 7058X75
 View more versions of this product
Catalog No. 23-750066
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Cytoplasmic counterstain for pap staining technique
    Type EA-36 Papanicolaou Staining Solution
    Quantity 1000mL
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback