EMD Millipore™ Histone H1, Calf Thymus, Protein

Manufacturer: EMD Millipore 3821505MG

 View more versions of this product

Catalog No. 38-215-05MG

  Check Availability
Add to Cart

    Specifications

    Preparation Method H2O or Neutral aqueous buffers
    Quantity 5mg
    Recombinant No
    Immunogen Histone H1
    Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up 6 months at −70°C.
    Regulatory Status RUO
    Format White lyophilized solid
    Formulation Sold on the basis of protein content.
    Host Species Bovine (calf thymus)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit