Learn More
MilliporeSigma™ HPTLC Silica Gel 60 Concentrating Zone Plates
allow users to quickly and easily apply any kind of sample, even large volumes of diluted samples.
$577.00 - $1633.00
Specifications
Backing Material Glass
Pore Size 60 Å
Type Concentrating Zone HPTLC Plate
For Use With (Application) Quick and Easy Sample AppliCatIon From Small to Large Volumes of Diluted Samples, Highly Facilitated Sample Loading, Better Resolution Due to Focused Bands
Thickness (Metric) 200 µm
View More Specs
Products 7
Catalog Number Mfr. No. Sorbent Dimensions (L x W) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Sorbent Dimensions (L x W) Quantity Price Quantity & Availability  
M1131870001
View Documents Promotion Details
MilliporeSigma™
1.13187.0001
Silica Gel 60 F254 with concentrating zone 5 x 10 cm 25/Pk.
Each for $577.00
Add to cart
M1137490001
View Documents Promotion Details
MilliporeSigma™
1.13749.0001
Silica Gel 60 with concentrating zone 20 x 10 cm 50/Pk.
Each for $1,207.00
Add to cart
M1137270001
View Documents Promotion Details
MilliporeSigma™
1.13727.0001
Silica Gel 60 F254 with concentrating zone 10 x 10 cm 25/Pk.
Each for $698.00
Add to cart
M1137280001
View Documents Promotion Details
MilliporeSigma™
1.13728.0001
Silica Gel 60 F254 with concentrating zone 20 x 10 cm 50/Pk.
Each for $1,212.00
Add to cart
M1137480001
View Documents Promotion Details
MilliporeSigma™
1.13748.0001
Silica Gel 60 with concentrating zone 10 x 10 cm 25/Pk.
Each for $616.00
Add to cart
M1150370001
View Documents Promotion Details
MilliporeSigma™
1.15037.0001
Silica Gel 60 Plates RP-18 concentrating zone 20 x 10 cm 25/Pk.
Each for $1,493.00
Add to cart
M1154980001
View Documents Promotion Details
MilliporeSigma™
1.15498.0001
Silica Gel 60 RP-18 F254 s with concentrating zone 20 x 10 cm 25/Pk.
Each for $1,633.00
Add to cart
Description
Description
Our concentrating zone plates are based on different adsorption properties of two adsorbents. The first is a large pore concentrating adsorbent where the samples are applied; the second is a selective layer for separation. Regardless of the spots’ shape, size, or position, the sample always concentrates as a narrow band where the two adsorbents overlap and where the separation starts. These plates allow users to quickly and easily apply any kind of sample, even large volumes of diluted samples.
Specifications
Specifications
Glass
Concentrating Zone HPTLC Plate
200 µm
60 Å
Quick and Easy Sample AppliCatIon From Small to Large Volumes of Diluted Samples, Highly Facilitated Sample Loading, Better Resolution Due to Focused Bands