EMD Millipore™ β-Interferon, Human, Recombinant Protein, CHO Cells

Manufacturer: EMD Millipore 4073182UG

 View more versions of this product

Catalog No. 40-731-82UG

  Check Availability
Add to Cart

    Specifications

    Preparation Method Reconstitute with sterile water to ≥100μg/mL, containing 0.1% HSA or BSA. Further dilute with aqueous buffer just prior to use.
    Purification Method SDS-PAGE
    Purity ≥98%
    Quantity 2μg
    Recombinant Yes
    Immunogen β-Interferon
    Storage Requirements Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 7 days at 4°C or for up to 4 months at −20°C. Avoid freeze/thaw cycles of solutions.
    Regulatory Status RUO
    Cell Type CHO Cells
    Format Lyophilized
    Formulation Lyophilized from 62mg/mL mannitol, 9.1mg/mL human albumin, 0.9mg/mL acetate, pH 7.0-7.2.
    Host Species Human

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit