Learn More
EMD Millipore™ β-Interferon, Human, Recombinant Protein, CHO Cells Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Millipore Sigma 40731810UG
 View more versions of this product
Catalog No. 40-731-810UG
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized
Reconstitute with sterile water to ≥100μg/mL, containing 0.1% HSA or BSA. Further dilute with aqueous buffer just prior to use.
≥98%
β-Interferon
RUO
Human
Lyophilized from 62mg/mL mannitol, 9.1mg/mL human albumin, 0.9mg/mL acetate, pH 7.0-7.2.
SDS-PAGE
10μg
Following reconstitution, refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 7 days at 4°C or for up to 4 months at −20°C. Avoid freeze/thaw cycles of solutions.
CHO Cells
Yes