Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ LiChroCART™ Column/LiChrospher™ RP-4 ADS Sorbent HPLC Cartridges, 25μm
HPLC cartridge
Supplier: MilliporeSigma™ 1.50380.0001
Description
LiChrospher™ ADS allows the fully automated, LC integrated sample preparation of hydrophobic, low molecular weight analytes from untreated samples like serum, milk, fermentation broth of food homogenates. The enrichment is based on two chromatographic processes: reversed phase and size exclusion chromatography since LiChrospher™ ADS sorbents are spherical silica gel particles (25μm) with DIOL modification on the exterior surface and alkyl modifications on the surface of the interior pores (ID: 6nm). Depending on the hydrophobicity of the analyte three types of precolumns are available: C-4, C-8 and C-18.Specifications
Cartridge | |
25 mm | |
2 to 7.5 | |
3/Pk. | |
C4 ADS | |
Mixed Mode | |
LiChroCART Column/LiChrospher RP-4 ADS Sorbent HPLC Cartridge |
2 mm | |
25 μm | |
60 Å | |
L26 | |
Silica | |
Reversed Phase, Ion-Pair, Size Exclusion |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title