Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ LiChroCART™ Column/LiChrospher™ RP-4 ADS Sorbent HPLC Cartridges, 25μm

HPLC cartridge

$929.50

Specifications

Length (Metric) 25 mm
Particle Size 25 μm
pH 2 to 7.5
Pore Size 60 Å
USP Type L26
View More Specs

Products 3
Catalog Number Mfr. No. Diameter (Metric) Quantity Type Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Quantity Type Price Quantity & Availability  
M1503800001
missing translation for 'viewDocuments'
MilliporeSigma™
1.50380.0001
2 mm 3/Pk. LiChroCART Column/LiChrospher RP-4 ADS Sorbent HPLC Cartridge
Each for $929.50
Add to cart
 
M1502060001
missing translation for 'viewDocuments'
MilliporeSigma™
1.50206.0001
4 mm 1/Pk. LiChroCART Column/LiChrospher RP-4 ADS Sorbent HPLC Cartridge Set This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
M1502080001
missing translation for 'viewDocuments'
MilliporeSigma™
1.50208.0001
4 mm 3/Pk. LiChroCART Column/LiChrospher RP-4 ADS Sorbent HPLC Cartridge This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

LiChrospher™ ADS allows the fully automated, LC integrated sample preparation of hydrophobic, low molecular weight analytes from untreated samples like serum, milk, fermentation broth of food homogenates. The enrichment is based on two chromatographic processes: reversed phase and size exclusion chromatography since LiChrospher™ ADS sorbents are spherical silica gel particles (25μm) with DIOL modification on the exterior surface and alkyl modifications on the surface of the interior pores (ID: 6nm). Depending on the hydrophobicity of the analyte three types of precolumns are available: C-4, C-8 and C-18.
Specifications

Specifications

25 mm
2 to 7.5
L26
Silica
Reversed Phase, Ion-Pair, Size Exclusion
25 μm
60 Å
C4 ADS
Mixed Mode
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit