Learn More
MilliporeSigma™ LiChroCART™ Column/Superspher™ Sorbent HPLC Cartridges, 4 micron
A high-performance spherical silica carrier with a mean particle size of 4μm provides an excellent pressure/separation performance ratio
Manufacturer:  MilliporeSigma™ 1.16009.0001
 View more versions of this product
Catalog No. M1160090001
Add to cart
Description
Description
  • Reduces solvent consumption
  • Allows handling of very small quantities with excellent sensitivity and resolution
  • High peak capacity
Specifications
Specifications
4 mm
2175 psi (150 bar) to 2900 psi (200 bar)
60 Å
700 m²/g
Silica
Normal Phase
HPLC Cartridge
250 mm
4 μm
1/Pk.
Silica
Normal Phase
Stainless Steel