Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
MilliporeSigma LiChrospher PAH HPLC Columns, 5µm
HPLC cartridge
$431.00 - $1049.00
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Quantity | Price | Quantity & Availability | |||
M1501490001
|
MilliporeSigma
1.50149.0001 |
4 mm | 250 mm | 1/Pk. |
Each for $1,049.00
|
N/A |
M1501480001
|
MilliporeSigma
1.50148.0001 |
4 mm | 4 mm | 10/Pk. |
Each for $431.00
|
N/A |
M1501560001
|
MilliporeSigma
1.50156.0001 |
3 mm | 250 mm | 1/Pk. |
Each for $976.50
|
N/A |
Description
LiChrospher PAH is a highly efficient and selective stationionary phase especially designed for the high resolution separation of 16 PAH (polycyclic aromatic hydrocarbons) according to EPA 610 and 550, plus benzo(e)pyrene and perylene. This sorbent is packed into LiChroCART HPLC guard columns, to be used in connection with LiChroCART HPLC cartridges, packed with LiChrospher PAH. The connection is being accomplished by use of the manu-CART NT cartrige holder (1.51486).Specifications
350 m²/g | |
HPLC Column |