Learn More
MilliporeSigma™ LiChrospher™ RP-18 ADS Cartridge Set
HPLC cartridge set, contains 1 LiChroCART™ 25-4 LiChrospher™ RP-18 ADS 1 manu-CART™
Supplier: MilliporeSigma™ 1.50187.0001
Description
LiChrospher™ ADS allows the fully automated, LC integrated sample preparation of hydrophobic, low molecular weight analytes from untreated samples like serum, milk, fermentation broth of food homogenates. The enrichment is based on two chromatographic processes: reversed phase and size exclusion chromatography since LiChrospher™ ADS sorbents are spherical silica gel particles (25μm) with DIOL modification on the exterior surface and alkyl modifications on the surface of the interior pores (ID: 6nm). Depending on the hydrophobicity of the analyte three types of precolumns are available: C-4, C-8 and C-18.Specifications
LiChrospher RP-18 ADS Cartridge Set |