Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Lumican, HisTag™, CF, Mouse, Recombinant Protein

Catalog No. 44031950UG Shop All MilliporeSigma Products
Click to view available options
Quantity:
50 μg

Catalog No. 44-031-950UG

Supplier: MilliporeSigma™ 44031950UG

Only null left
Add to Cart
Add to Cart
Recombinant, murine lumican, consisting of amino acids 19-338, fused at the C-terminus to a His·Tag™ sequence, and expressed in NS0-derived mouse myeloma cells. Lumincan a 40kDa member of the small leucine-rich repeat proteoglycans (SLRPs) family/Class II subfamily of proteins. It is known for its ability to modulate collagen fibrillogenesis and role in cell proliferation regulation.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.2μM filtered PBS.
Preparation Method Reconstitute to 200μg/mL with sterile PBS.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen Lumican
Storage Requirements Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C.
Regulatory Status RUO
Without Additives Carrier-Free
Conjugate His-Tag
Species Mouse
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.