Learn More
MilliporeSigma™ M Air T™ Agar Cassettes
Ready-to-use cassettes, prefilled with gamma-sterilized medium
Specifications
For Use With (Equipment) | M air T Air Testing System |
---|---|
Certifications/Compliance | USP |
Catalog Number | Mfr. No. | Type | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | For Use With (Application) | Price | Quantity & Availability | ||||
ATSMTTD60
|
MilliporeSigma™
ATSMTTD60 |
M air T Cassettes | For the total count of airborne microorganisms in pharmaceutical clean rooms, hospitals, food and beverage applications | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
ATSMC0114
|
MilliporeSigma™
ATSMC0114 |
M air T Empty Cassettes, Sterile | For the manual production of Culture Media, for special applications in the microbiological air monitoring, pharmaceutical products, environmental analysis | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Fast and reliable results with maximum recovery
- Large agar amount and unique micro-perforated sieve allow air sampling of up to 1 cubic meter
- Unit-to-unit consistency of agar surface shape
- D
- Reproducible results
Compliance
USP
Specifications
M air T Air Testing System | |
USP |