Learn More
MilliporeSigma™ Malachite Green Oxalate Dye
For microscopy and for microbiology. CAS 2437-29-8, EC Number 219-441-7, chemical formula C₄₆H50N₄ * 2 C2HO₄ * C2H2O₄.
Manufacturer:  MilliporeSigma™ 1.01398.0100
 View more versions of this product
Catalog No. M1013980100
Add to cart
Description
Description
MALACHITE GREEN OXALATE (C.I. 42000) is a dry dye that is used for the preparation of a staining solution that can be used for a spore staining according to Rakette. It is possible to use Malachite green dye for differential spore staining in bacteriological samples of human origin, Dye is used in a hot staining method to stain specifically spores. Counterstain with a red dye for better visualization. The dye has a constant dye content and a reliable high quality and thus, is registered and certified as IVD and CE product, one of the major precondition for a reproducible diagnostic staining.
Specifications
Specifications
100g
Malachite Green Oxalate
Green