Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma MAP Kinase 1, HisTag, Human, Recombinant Protein, E. coli
Manufacturer: MilliporeSigma 4548492.5UG
Description
Recombinant, human MAP Kinase 1 fused to a His·Tag sequence and expressed in E. coli. Highly active form produced by phosphorylation of the purified MAP Kinase in vitro with MEK1. Suitable for labeling MAP Kinase substrates.Specifications
Liquid | |
SDS-PAGE | |
20.5μg | |
MAP Kinase 1 | |
RUO | |
Human |
In 100mM NaCl, 50mM Tris-HCl, 1mM DTT, 50% glycerol, pH 8.5. | |
≥95% | |
E. coli | |
Following initial thaw, aliquot and freeze (−70°C). | |
His·Tag™ | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title