Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MAP Kinase 2, HisTag™, Human, Recombinant Protein, E. coli

Catalog No. 4548545UG
Click to view available options
Quantity:
5 μg
Recombinant, human MAP Kinase 2 fused to a His·Tag™ sequence and expressed in E. coli. Highly active form produced by phosphorylation of the purified MAP Kinase in vitro with MEK1. Suitable for labeling MAP Kinase substrates.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 150mM NaCl, 50mM Tris, 1mM DTT, 50% glycerol, pH 8.5.
Purification Method SDS-PAGE
Quantity 5 μg
Source E. coli
Immunogen MAP Kinase 2
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate His-Tag
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.