Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ MQuant™ pH Indicator Litmus Papers
Measure pH swiftly without using instruments
$80.82
Specifications
For Use With (Application) | For long term storage, since they are protected against external influences, pH indicator papers are manufactured by impregnating high quality filter paper with indicator or mixed indicator solutions |
---|---|
Includes | With color scale |
Quantity | 3 Rolls |
Storage Requirements | 10°C to 25°C |
Type | Litmus Paper |
Includes: Color scale
Catalog Number | Mfr. No. | Color Indicator | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color Indicator | Price | Quantity & Availability | |||||
M1094860003
|
MilliporeSigma™
1.09486.0003 |
Blue |
Each of 1 for $80.82
|
|
Description
Suited for all media in environmental analysis and industrial in-process controls.
- More precise evaluation due to differentiated color graduations
- Biologically degradable polyester foil and low reagent content in reaction zones support problem-free waste disposal
- Range: pH <7, red; pH >7, blue
- M1094860003: blue
- M1095180003: neutral
- M1094890003: red
- M1094860003 and M1094890003 are Reag. Ph. Eur grade
Order Info
Three rolls of 4.8m
Specifications
For long term storage, since they are protected against external influences, pH indicator papers are manufactured by impregnating high quality filter paper with indicator or mixed indicator solutions | |
3 Rolls | |
Litmus Paper |
With color scale | |
10°C to 25°C |
Safety and Handling
ShelfLife : 3 to 5 Years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title