Learn More
MilliporeSigma™ MColortest™ Ozone Test Kits
This test measures the content of ozone in drinking water, wastewater, process water and disinfecting solutions. The concentration can be measured semiquantitatively by using the colour comparator.
Manufacturer:  MilliporeSigma™ 1.18758.0001
 View more versions of this product
Catalog No. M1187580001
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
0.15 - 10mg/L O3
Ozone Test Kit
Colorimetric, DPD, with color-disk comparator 0.15 - 0.35 - 0.5 - 0.7 - 1.4 - 2.7 - 5.0 - 7.0 - 10 mg/l O3 MColortest