Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma MEK1, GST-Fusion, Human, Recombinant Protein, E. coli
Manufacturer: MilliporeSigma 47570210UG
Description
Recombinant, human MEK1 fused to GST at the N-terminus and expressed in E. coli. MEK1 is a member of a dual-specificity protein kinase family that phosphorylates threonine and tyrosine residues on ERK 1 and 2.Specifications
Concentration on Label | |
In 270mM sucrose, 150mM NaCl, 50mM Tris-HCl, 1mM DTT, 0.1mM EGTA, pH 7.5. | |
≥80% | |
E. coli | |
Following initial thaw, aliquot and freeze (−70°C). | |
GST-Fusion | |
Yes |
Liquid | |
SDS-PAGE | |
10μg | |
MEK1 | |
RUO | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title