Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MF-Millipore™ Mixed Cellulose Ester Membranes: 0.80µm Pore Size Available on GSA/VA Contract for Federal Government customers only.

Designed for air monitoring, bioassays, dairy microbiology, particle monitoring and particle removal. MF-Millipore™ Mixed Cellulose Ester Membranes, 0.80µm pore size are available in a range of diameters. Composed of mixed cellulose membrane in black or white with a plain or gridded surface.

Manufacturer:  MilliporeSigma™ AAWP03700

 View more versions of this product

Catalog No. AAWP03700


Add to cart

Description

Description

  • Hydrophilic, 150µm thick membranes with a plain or gridded surface
  • Composed of a biologically inert mixture of cellulose acetate and cellulose nitrate
  • Widely used with fuel and aviation turbine fluid contamination SDI, particles and insoluble contaminants
  • White membranes
    Ideal for air monitoring, bioassays, dairy microbiology, particle monitoring, and particle removal
  • Black membranes
    Ideal for Air monitoring, fluorescent assays, and particle monitoring

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

≥1.0 bar, air with water
37mm
Hydrophilic
150µm
Mixed Cellulose Esters (MCE)
Plain
White
190mL/min/cm sq. (Liquid), 16L/min/cm sq. (Gas)
100/pk.
For air monitoring, bioassays, dairy microbiology, particle monitoring, and particle removal
0.8μm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit