Learn More
MilliporeSigma™ Microbiological Analysis Membrane Filters
Optimized for MF method microbiological analysis of water or other liquids
Supplier:  MilliporeSigma™ GSWP047S0
 View more versions of this product
Catalog No. GSWP047S0
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.
Description
Description
  • MF-Type (mixed cellulose esters) filter membranes
  • 100 Sterile pack: 10 resealable envelopes containing 10 filters and 10 pads, sterile 
Compliance
Certified to meet Standard Methods for the Examination of Water and Wastewater requirements.
Specifications
Specifications
Certified to meet standard methods for the examination of Water and wastewater requirements
0.22 μm
MF method microbiological analysis of water or other liquids
SO Pak Membrane Filter
4.7 cm
White
Sterile
Plain
1.85 in.