Learn More
MilliporeSigma™ Microfil™ S Filtration Devices
Fast, dependable alternative to conventional multi-step methods
Supplier: MilliporeSigma™ MVGSWGS24
Description
Devices consist of a funnel, protective lid, and filtration support so routine microbiological testing is fast and easy.
- Ensure consistent recovery of microorganisms
- Smooth, hydrophobic funnel surface repels aqueous sample residues to ensure that microorganisms are collected on the membrane, not on funnel walls
- Convex support shape eliminates pleating and distortion of membrane due to expansion
- Three membrane selections offer versatility for your specific application
- Presterilized, individually packaged No membrane handling prior to filtering
- No filter holders, funnels and attachments to assemble and sterilize
- No sterilizing the filter support after each filtration Protective lid reduces external contamination risk Pre-assembled to speed sampling and shorten preparation Easy-to-remove funnel and membrane reduce test time
Compliance
Comply with USP/EP Guidelines. Certificate of Quality demonstrates that the device has been quality-control tested with Pharmacopoeia organisms and demonstrates acceptable growth.
Specifications
EMD Millipore Microfil S Filtration Devices | |
Mixed Cellulose Ester | |
0.22 μm |