Learn More
MilliporeSigma™ Millex™ Filters: Inlet and Outlet, 1/8 NPT Available on GSA/VA Contract for Federal Government customers only.
Protect vacuum sources from contamination and moisture damage
Manufacturer:  MilliporeSigma™ SLFG75010
Catalog No. SLFG75010
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • With hydrophobic PTFE membranes
  • Polypropylene housing (except for SLFG-025-LS and -50, which have PVC housing)
  • Variety of inlet/outlet combinations, including stepped hose barb and 1/8 NPT
  • Nonsterile, autoclavable, bidirectional
Applications:
  • Protect vacuum sources from contamination and moisture damage
  • Sterilize ambient air entering fermenters
  • Filter incubator gas to protect against contamination
  • Filter nitrogen and air used to dry liquid samples
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Non-sterile
0.2μm
Polypropylene
Protect vacuum sources from contamination and moisture damage; Sterilize ambient air entering fermenters; Filter incubator gas to protect against contamination; Filter nitrogen and air used to dry liquid samples
50mm
PTFE
Yes