Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
MilliporeSigma™ Millex-MP Filter Unit Available on GSA/VA Contract for Federal Government customers only.
A 25mm diameter sterile syringe filter with a 0.22µm pore size Polyethersulfone membrane. Comes in a pack of 50.
Manufacturer:  MilliporeSigma™ SLMP025SS
Catalog No. SLMP025SS
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0.22μm
50 Pack
Clear
Low protein binding, sterilize tissue culture media, protein solutions, and other aqueous solutions
0.98 in.
113°F
100mL
Hydrophilic Polyethersulfone
Sterile
2.5cm
Female luer lok/male luer slip
3.9cm2
2.5cm
5.2 bar
45°C
<100μL
Biosart™ 100 Monitor