Learn More
MilliporeSigma™ Millex™ Nonsterile Syringe Filters with PE Housing - Hydrophobic PTFE
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Manufacturer:  MilliporeSigma™ SLFG025NK
 View more versions of this product
Catalog No. SLFG025NK
Add to cart
Description
Description
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
  • Compatible with more aggressive organic solutions and high temperature conditions
Specifications
Specifications
Non-sterile
25mm
Polyethylene
1000/Pk.
Syringe Filter
Polytetrafluoroethylene (PTFE)
0.2μm