Learn More
MilliporeSigma™ Millex™ Nonsterile Syringe Filters with PE Housing - Nylon Available on GSA/VA Contract for Federal Government customers only.
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
$226.00 - $2180.00
Specifications
Type Syringe Filter
Material Nylon (NYL)
View More Specs
Products 6
Catalog Number Mfr. No. Diameter (Metric) Pore Size Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Pore Size Quantity Price Quantity & Availability  
SLGN033NK Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
SLGN033NK
33mm 0.2μm 1000/Pk.
Pack of 1000 for $2,180.00
Add to cart
SLGN033NS
View Documents
MilliporeSigma™
SLGN033NS
33mm 0.2μm 50/Pk.
Pack of 50 for $226.00
Add to cart
SLGN033NB Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
SLGN033NB
33mm 0.2μm 250/Pk.
Pack of 250 for $909.00
Add to cart
SLHN033NB
View Documents
MilliporeSigma™
SLHN033NB
33mm 0.45μm 250/pk.
Pack of 250 for $909.00
Add to cart
SLHN033NK Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
SLHN033NK
33mm 0.45μm 1000/Pk.
Pack of 1000 for $2,180.00
Add to cart
SLHN033NS Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
SLHN033NS
33mm 0.45μm 50/Pk.
Pack of 50 for $226.00
Add to cart
Description
Description
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
  • Broad chemical compatibility Choose for filtering aqueous and organic solutions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Syringe Filter
Nylon (NYL)