Learn More
MilliporeSigma™ Millex™ Nonsterile Syringe Filters with PE Housing - PVDF Membrane - 25mm Diameter
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Manufacturer:  MilliporeSigma™ SLHV025NK
 View more versions of this product
Catalog No. SLHV025NK
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • With female Luer-Lok™ inlet and male Luer slip outlet
  • Lowest protein binding
  • Use for aqueous and mild organic solutions
Specifications
Specifications
Syringe Filter
PVDF Membrane
Polyethylene
25mm
Use for aqueous and mild organic solutions
0.45μm