Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Millicup™ Self-Contained, Disposable, Vacuum Bottle-Top Filter for HPLC
Non sterile Universal bottletop vacuum filter
$203.50 - $221.00
Specifications
Type | Bottle Top Filter |
---|---|
For Use With (Application) | HPLC |
Height (English) | 4.7 in. |
Height (Metric) | 12 cm |
Pore Size | 0.45 μm |
Catalog Number | Mfr. No. | Membrane Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Membrane Material | Price | Quantity & Availability | |||||
SJHVM4710
|
MilliporeSigma™
SJHVM4710 |
Durapore PVDF |
Pack of 10 for $215.00
|
|
|||||
SJFHM4710
|
MilliporeSigma™
SJFHM4710 |
Hydrophobic PTFE |
Pack of 10 for $221.00
|
|
|||||
SJLHM4710
|
MilliporeSigma™
SJLHM4710 |
Hydrophilic PTFE |
Pack of 10 for $203.50
|
|
|||||
Description
The Millicup vacuum-driven bottletop filter unit is a disposable device that provides a fast, effective, and safe means to filter mobile phases and running buffers used in instrumentation analyses (for example, HPLC). The Millicup vacuum-driven bottletop filter unit eliminates the risk of contaminant carry-over from solvent to solvent. Unlike reusable filter holders that must be washed and assembled before each use, Millicup units require no assembly or clean-up. If accidentally dropped, the Millicup vacuum-driven bottletop filter unit will not break, ensuring a safe environment in which to filter solvents and other hazardous solutions.Specifications
Bottle Top Filter | |
4.7 in. | |
0.45 μm |
HPLC | |
12 cm | |
8.4 cm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title