Learn More
MilliporeSigma Millipak Disposable Filter Units
Removes particles and microorganisms from liquids and gases with pump-driven filter. Millipak Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.
$113.90 - $330.00
Specifications
Diameter (English) | 3 in. |
---|---|
Diameter (Metric) | 7.6 cm |
Material | Polycarbonate |
Membrane Material | PVDF |
Max. Inlet Pressure | 75 psi at 25°C |
Catalog Number | Mfr. No. | Connectors | Flow Rate (Metric) | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Connectors | Flow Rate (Metric) | Volume (Metric) | Price | Quantity & Availability | |||
MPGL02GF2
|
MilliporeSigma
MPGL02GF2 |
0.75 in./19.05mm sanitary flange inlet and outlet | 1.5 L/min. at 1.75 bar | 10 L |
Pack of 2 for $160.90
|
|
MPGL02GH2
|
MilliporeSigma
MPGL02GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 1.5 L/min. at 1.75 bar | 20 L |
Each for $159.00
|
|
MPGL04GH2
|
MilliporeSigma
MPGL04GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 2.8 L/min. at 1.75 bar | 20 L |
Pack of 2 for $189.60
|
|
MPGL04GF2
|
MilliporeSigma
MPGL04GF2 |
0.75 in./19.05mm sanitary flange inlet and outlet | 2.8 L/min. at 1.75 bar | 20 L |
Pack of 2 for $192.30
|
|
MPGL06CB1
|
MilliporeSigma
MPGL06CB1 |
0.25 in./6.35mm NPTM inlet, 0.25 in./6.35mm stepped hose barb with bell outlet | 7.6 L/min. at 2 bar | 30 L |
Each for $113.90
|
|
MPGL06GF2
|
MilliporeSigma
MPGL06GF2 |
0.75 in./19.05mm sanitary flange inlet and outlet | 7.6 L/min. at 2 bar | 30 L |
Pack of 2 for $212.00
|
|
MPHL02GF2
|
MilliporeSigma
MPHL02GF2 |
0.75 in./19.05mm sanitary flange inlet and outlet | 6.4 L/min. at 2.45 bar | 10 L |
Pack of 2 for $155.00
|
|
MPGL06GH2
![]() |
MilliporeSigma
MPGL06GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 3.8 L/min. at 1.75 bar | 30 L |
Each for $212.00
|
|
MPHL06GF2
|
MilliporeSigma
MPHL06GF2 |
0.75 in./19.05mm sanitary flange inlet and outlet | 10.5 L/min. at 2.45 bar | 30 L |
Pack of 2 for $224.50
|
|
MPHL06GH2
|
MilliporeSigma
MPHL06GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 10.5 L/min. at 2.45 bar | 30 L |
Each for $221.50
|
|
MPHL04GH2
|
MilliporeSigma
MPHL04GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 8.5 L/min. at 2.45 bar | 20 L |
Each for $177.70
|
|
MPHL02GH2
|
MilliporeSigma
MPHL02GH2 |
0.25 in. /6.35mm stepped hose barb inlet and outlet with bell | 6.4 L/min. at 2.45 bar | 10 L |
Each for $157.80
|
|
MPSL06CB1
|
MilliporeSigma
MPSL06CB1 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 7.6 L/min. at 700mbar | 30 L |
Each for $134.30
|
|
MPSL06CL1
|
MilliporeSigma
MPSL06CL1 |
1.5 in./38.1mm sanitary flange inlet, 0.25 in./6.35mm stepped hose barb outlet | 7.6 L/min. at 700mbar | 30 L |
Each for $134.30
|
|
MPVL02GH2
|
MilliporeSigma
MPVL02GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 0.7 L/min. at 1.75 bar | 10 L |
Each for $162.70
|
|
MPVL06GH2
|
MilliporeSigma
MPVL06GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 1.9 L/min. at 1.75 bar | 30 L |
Each for $233.50
|
|
MPVL04GH2
|
MilliporeSigma
MPVL04GH2 |
0.25 in./6.35mm stepped hose barb inlet and outlet with bell | 1.1 L/min. at 1.75 bar | 20 L |
Each for $196.60
|
|
MPGL04GK2
![]() |
MilliporeSigma
MPGL04GK2 |
0.25 in./6.35mm NPTM inlet, 0.25 in./6.35mm stepped hose barb with bell outlet | 2.8 L/min. at 1.75 bar | 20 L |
Pack of 2 for $330.00
|
|
Description
Removes particles and microorganisms from liquids and gases with pump-driven filter. Millipak Disposable Filter Units provide sterile filtration of parenterals, diagnostics, buffers, media, and biologicals.
Unique, stacked-disc design allows minimal hold-up volume and eliminates the particle-generating, woven, downstream support found in pleated cartridges.
- Sterile filtration of parenterals, diagnostics, buffers, media, and biologicals
- Point-of-use filter for high-purity water
- Ideally suited for high value-added applications
- Durapore PVDF membranes provide high flowrates and throughputs, low extractables, broad chemical compatibility, and the lowest protein binding of any membrane available Three different stack sizes allow optimum flexibility for easy scale-up and system sizing
- Presterilized units are gamma irradiated
- Autoclavable up to three times Each unit is labeled with catalog number, lot number, pore size and serial number
- 100% integrity tested to guarantee integrity
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
3 in. | |
Polycarbonate | |
75 psi at 25°C |
7.6 cm | |
PVDF | |
Filter Unit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.