Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Millistak+™ Pod Disposable Depth Filter System
Ideal for primary and secondary clarification in lab, pilot and process-scale applications
Supplier: MilliporeSigma™ MDE60054H1
Description
- Filter Material: cellulose
- Configurations: 0.11, 0.77, or 1.4 m2 filtration area
- Low hold-up volume for greater product yield
- Broad range of 23 media types offered in single and multilayer products
- Flexible, modular format offers scalability from 5 to 12,000L or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Disposable Pod system protects operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Specifications
Single-use | |
Cellulose, Inorganic Filter Aid | |
Clarification of serum, plasma, vaccines, cell culture or other fluids | |
0.5 in. hose barb | |
1 Ea. | |
5 to 12,000 L | |
21.59 cm | |
13.46 cm |
Glass Fiber-reinforced Polypropylene | |
0.054 m2 | |
DE60 | |
50 psi at 25°C, 15 psi at 80°C | |
Millistak+ Pod Depth Filter | |
8.5 in. | |
5.3 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction