Learn More
MilliporeSigma™ Montage™ Gel Extraction Kit
Elutes DNA from agarose gels in 10 minutes
Supplier:  MilliporeSigma™ LSKGEL050
Catalog No. LSKGEL050
Add to Cart
Includes: 50 gel nebulizers, 50 Ultrafree-MC 0.45μm separators, 50 microcentrifuge vials, 500mL stock of 50X modified TAE buffer
Description
Description
  • Typically recovers >70% of 100-1000 bp DNA from standard agarose gels
  • Converts agarose to a spray from which DNA is separated via a 0.45μm Ultrafree™-MC filter
  • Prepares gel-purified PCR products for sequencing or cloning without need for further purification
Specifications
Specifications
50 Gel nebulizers, 50 Ultrafree™-MC 0.45μm separators, 50 microcentrifuge vials, 500mL stock of 50X modified TAE Buffer
50 Pack
Product Suggestions
Product Suggestions