Learn More
EMD Millipore™ MQuant™ Ammonium Test Strips Available on GSA/VA Contract for Federal Government customers only.
High-tech, fast and simple means for semiquantitative testing
Manufacturer:  EMD Millipore™ 1.10024.0001
Catalog No. M1100240001
Add to cart
Includes: Reagent
Description
Description
Uses Nessler method
  • Small and easy to handle for on-site use
  • Fast results
  • Ideal for screening
  • PET film and low reagent content for easier disposal
  • Graduation: 10-30-60-100-200-400mg/L
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
The test is suitable for determination potentially harmful pollution in surface and ground water, which are caused by ammonium compounds, in agriculture the test can be used to measure the ammonium nitrogen content of various types of manure and compost
2 bottles of reagent NH4-1, 1 test vessel
15° to 25°C
MQuant™ Ammonium Test Strip
10-30-60-100-200-400mg/L
Each
Neuler