Learn More
EMD Millipore™ MQuant™ Chloride Test Strips Available on GSA/VA Contract for Federal Government customers only.
To measure chloride ions in all natural waters
Manufacturer:  EMD Millipore™ 1.10079.0001
Catalog No. M1100790001
Add to cart
Includes: Tube containing 100 strips
Description
Description
  • Small and easy to handle
  • Flexible and universal application
  • Silver Chromate reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable
  • Color-scale graduation: 500 - 1000 - 1500 - 2000 - 3000mg/L
Reclose the tube containing the test strips immediately after use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Measuring chloride ions in all natural water
500-1000-1500-2000-3000mg/L
2° to 8°C
Colorimeter
500 to 3000mg/L
Test Strip